missing translation for 'onlineSavingsMsg'
Learn More

Fetuin-A Rabbit anti-Human, Clone: 10000, Proteintech

Product Code. 17639796 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
Product Code. Quantity unitSize
17639796 100 μL 100µL
17649796 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 17639796

Brand: Proteintech 807931RR100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Antibody

Fetuin-A antibody apecifically detetects Fetuin-A antigen in Human Samples. It is validated for Western Blot.

Specifications

Antigen Fetuin-A
Applications Western Blot
Clone 10000
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
Gene Accession No. BC048198
Gene Symbols AHSG
Host Species Rabbit
Immunogen Fetuin-A fusion protein Ag9853
Purification Method Protein A purification
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 197
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.