missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferroportin/SLC40A1 Antibody (1308C) - BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
1435.00 NOK - 4465.00 NOK
Specifications
| Antigen | Ferroportin/SLC40A1 |
|---|---|
| Clone | 1308C |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 0.25 μg/mL, Immunohistochemistry 1 μg/mL, Immunohistochemistry-Paraffin 1 μg/mL |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18230209
|
Novus Biologicals
NBP2-75923SS |
0.025 mg |
1435.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18041196
|
Novus Biologicals
NBP2-75923 |
0.1 mg |
4465.00 NOK
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ferroportin/SLC40A1 Monoclonal specifically detects Ferroportin/SLC40A1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Ferroportin/SLC40A1 | |
| 1.0 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Ferroportin-1, FPN1, FPN1IREG1ferroportin 1, HFE4, HFE4ferroportin-1, IREG1, iron regulated gene 1, Iron-regulated transporter 1, member 3, MST079, MSTP079, MTP1, putative ferroportin 1 variant IIIB, SLC11A3, SLC11A3iron regulated gene 1, solute carrier family 11 (proton-coupled divalent metal ion transporters), solute carrier family 11 (proton-coupled divalent metal ion transporters), member 3, solute carrier family 40 (iron-regulated transporter), member 1, solute carrier family 40 member 1 | |
| SLC40A1 | |
| IgG | |
| Protein A or G purified | |
| 62.5 kDa |
| 1308C | |
| Flow Cytometry 0.25 μg/mL, Immunohistochemistry 1 μg/mL, Immunohistochemistry-Paraffin 1 μg/mL | |
| Monoclonal | |
| Purified | |
| Cancer, Lipid and Metabolism, Signal Transduction | |
| PBS with 0.02% Sodium Azide | |
| 30061.0 | |
| This Ferroportin/SLC40A1 Antibody (1308C) was developed against a synthetic peptide made to an internal portion of human Ferroportin 1 (within residues 250-320). [UniProt# Q9NP59] | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title