Learn More
Thermo Scientific™ TopVision Agarose
Product Code. 10588360 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10588360 500 g 500g
10784691 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 10588360 Supplier Thermo Scientific™ Supplier No. R0492
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
 Restricted Item
In Stock
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
TopVision Agarose is highly purified agarose suitable for analytical and preparative electrophoresis of nucleic acids.
Thermo Scientific TopVision Agarose provides optimal concentration between 0.7 to 2% in all typical buffer systems. It is a highly purified agarose with very low EEO values certified by strict quality control test procedures.
TRUSTED_SUSTAINABILITY
Specifications
Separation Range 100 to >30000 bp
Melting Point 88 °C
Quantity 500 g
Validated Application Gel Electrophoresis
Product Line TopVision
Product Type Agarose Gel