Learn More
Thermo Scientific™ NoLimits 35 bp DNA Fragment
Shop All Thermo Scientific Products
Description
NoLimits is a collection of 36 highly-purified individual DNA fragments ranging from 10 to 20,000 bp. Unlike other vendors, where custom standards are produced by PCR, Thermo Scientific NoLimits DNA fragments are produced using specifically designed plasmids purified by patented technology.
Plasmid DNA is digested with Thermo Scientific Restriction Enzymes, and the individual DNA fragments are chromatography-purified from the digestion mixture. As a result, we offer exceptionally pure DNA fragments that are free of any truncated or degraded molecules. Our custom DNA fragments are the best choice for applications, ranging from electrophoresis (both gel and capillary) to HPLC and beyond.
Highlights
- Chromatography-purified individual DNA fragments
- Broad range: 10 to 20,000 bp
- Sharp peaks during capillary electrophoresis, sharp bands during gel electrophoresis
- Precise DNA concentration
Specifikationer
Specifikationer
| Gel Type | Agarose |
| Product Type | DNA Fragment |
| Ready to Load | No |
| Size Range | 10 to 20000 bp |
| Gel Compatibility | Agarose Gels |
| Green Features | Sustainable packaging |
| Product Line | NoLimits |
| Quantity | 10 μg |
For Research Use Only. Not for use in diagnostic procedures.
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.