Learn More
Thermo Scientific™ Maxima™ Hot Start Green 2X PCR Master Mix
Shop All Thermo Scientific ProductsDescription
Maxima Hot Start Green PCR Master Mix (2X) is a ready-to-use solution containing Maxima Hot Start Taq DNA Polymerase, optimized hot start PCR buffer, Mg2+, and dNTPs. The master mix is supplemented with two tracking dyes and a density reagent that allows for direct loading of PCR products on a gel.
The dyes in the master mix do not interfere with PCR performance and are compatible with downstream applications such as DNA sequencing, ligation and restriction digestion. The master mix retains all features of Maxima Hot Start Taq DNA Polymerase. It is capable of amplification of up to 3 kb from genomic DNA.
- Convenient—Maxima Hot Start Taq DNA Polymerase in a ready-to-use mix
- Hot start PCR—reaction set-up at room temperature
- Four minute activation time
- Direct loading of PCR product on gels
- Highs specificity and sensitivity of PCR
Recommended for:
High throughput hot start PCR; RT-PCR; Highly specific amplification of complex genomic and cDNA templates; Amplification of low copy DNA targets; Generation of PCR products for TA cloning
Specifications
Specifications
| Product Type | Fermentas™ Maxima™ Hot Start |
| Quantity | 10 x 1.25mL |
| Sufficient For | 500 reactions of 50 µl |
| For Use With (Application) | Standard PCR, RT-PCR |
Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.