missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BCIP-T

Product Code. 10765081 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10765081 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10765081 Supplier Thermo Scientific™ Supplier No. R0821

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A chromogenic substrate for alkaline phosphatase used in conjunction with the oxidant NBT (nitro blue tetrazolium, †R0841) to enhance blue color development.

Thermo Scientific™ Fermentas BCIP-T (5-bromo-4-chloro-3-indolyl phosphate, p-toluidine salt) is a chromogenic substrate for alkaline phosphatase used in conjunction with the oxidant NBT (nitro blue tetrazolium, †R0841) to enhance blue color development.

Alkaline phosphatase catalyses the removal of a phosphate group from BCIP-T, generating a product that oxidizes and dimerizes to dibromodichloro indigo. The reducing equivalents produced during the dimerization reaction reduce NBT to an insoluble purple dye, diformazan (1)

Recommended for:

Detection of alkaline phosphatase activity in:

  • Southern blotting
  • Western blotting
  • Northern blotting
  • dot/slot blots
  • plaque and colony screening

Note:

Preparation of a 50mg/mL stock solution in dimethylformamide is recommended.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Detection of alkaline phosphatase activity in:; Southern blotting Western blotting Northern blotting dot/slot blots plaque and colony screening;
Product Type BCIP-T
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.