Learn More
Thermo Scientific™ TAE Buffer (Tris-acetate-EDTA) (50X)
Product Code. 10399519 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10399519 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10399519 Supplier Thermo Scientific™ Supplier No. B49
  • ONLINE EXCLUSIVE PRICE
    866.00 NOK
    805.65 NOK / 1 L
    Save 60.35 NOK
    6% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Optimize electrophoresis of both genomic and large supercoiled DNA in agarose and polyacrylamide gels with Thermo Scientific™ 50X TAE Buffer (Tris-acetate-EDTA).
Use the Thermo Scientific 50X TAE Buffer (Tris-acetate-EDTA) for electrophoresis of nucleic acids in agarose and polyacrylamide gels. You can use this buffer for both genomic and large supercoiled DNA, and you can also use this as both a running and a gel preparation buffer.
Usage Recommendations
  • Use fresh 1X TAE both for the gel and for the electrophoresis run
  • To prepare 1X TAE buffer add 20ml of 50X TAE buffer to 980ml of deionized water and mix well
Quality Control
  • The absence of endo-, exodeoxyribonucleases and ribonucleases confirmed by appropriate quality tests
1X Composition
  • 40mM Tris
  • 20mM acetic acid
  • 1mM EDTA
  • pHof 50X TAE: 8.4
Recommended for:
Electrophoresis of nucleic acids in agarose and polyacrylamide gels; Used both as a running buffer and as a gel preparation buffer; Filtered through a 0.22μm membrane; Recommended for resolution of RNA and DNA fragments l.
Note:
TAE buffer has a relatively low buffering capacity, therefore periodic replacement of the buffer during prolonged electrophoresis is recommended.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 50X
Quantity 1 L
Product Type TAE Buffer
compliance-icons
Product Identifier
  • TAE Buffer (Tris-acetate-EDTA) (50X)
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: acetic acid