Learn More
Thermo Scientific™ 5-Fluoroorotic Acid
Product Code. 10619920 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10619920 5 g 5g
10609920 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10619920 Supplier Thermo Scientific™ Supplier No. R0812
  • ONLINE EXCLUSIVE PRICE
    3605.00 NOK
    3028.65 NOK / 5g
    Save 576.35 NOK
    15% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Detect expression of the URA3 gene, which encodes orotine-5'-monophosphate (OMP) dicarboxylase with Thermo Scientific™ 5-Fluoroorotic Acid, used in yeast molecular genetics.
5-Fluoroorotic Acid is a yeast with an active URA3 gene (Ura+) converts 5-FOA to fluorodeoxyuridine, which is toxic to cells (see the scheme below). Yeast strains carrying a mutation in the URA3 gene grow in the presence of 5-FOA if the media is supplemented with uracil. 5-FOA is frequently used in experiments with Saccharomyces cerevisiae (URA3), Schizosaccharomyces pombe (URA4 and URA5), Candida albicans (URA3), and E. coli (pyrF).
Yeast with an active URA3 gene (Ura+) converts 5-FOA to fluorodeoxyuridine, which is toxic to cells (see the scheme below). Yeast strains carrying a mutation in the URA3 gene grow in the presence of 5-FOA if the media is supplemented with uracil. 5-FOA is frequently used in experiments with Saccharomyces cerevisiae (URA3), Schizosaccharomyces pombe (URA4 and URA5), Candida albicans (URA3) and E.coli (pyrF).
Recommended for:
Detection of URA3 gene expression in yeast; Construction of yeast 2-hybrid libraries (1); Other areas of yeast genetics including gene replacement, analysis of transposable genetic elements and identification of gene mutations.
Note:
Low solubility in water. Solubility in dimethylsulfoxide is greater than 150mg/mL. Preparation of a 100mg/mL solution in dime-thylsulfoxide is recommended.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Fluoroorotic Acid
Product Type 5-Fluoroorotic Acid
Quantity 5 g
Form Powder