Learn More
BRAND™ FEP Filling Tube
Product Code. 10636863 Shop All Brand Products
Change view
Click to view available options
Diameter (Metric) Outer:
6.0 mm
7.6 mm
Overall Length (Metric):
1.5 m
10 m
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Overall Length (Metric) unitSize
10636863 6.0 mm 1.5 m Each
10151581 7.6 mm 10 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10636863 Supplier BRAND™ Supplier No. 704209
  • ONLINE EXCLUSIVE PRICE
    336.00 NOK
    318.68 NOK / Pack of 1
    Save 17.32 NOK
    5% Off
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
Manufactured from FEP. BRAND™ FEP Filling Tube dispenses accurate volumes directly from drums and bulk refills. For use with Dispensette™III/Organic.
  • Manufactured from FEP.
  • Dispenses accurate volumes directly from drums and bulk refills.
  • For use with Dispensette™III/Organic
TRUSTED_SUSTAINABILITY
Specifications
Material FEP
Diameter (Metric) Outer 6.0 mm
Overall Length (Metric) 1.5 m
For Use With Dispensette™III/Organic