Learn More
Fenuron, TRC
Specifications
Specifications
| Chemical Name or Material | Fenuron |
| CAS | 101-42-8 |
| Molecular Formula | C9 H12 N2 O |
| Quantity | 1 g |
| Synonym | Urea, N,N-dimethyl-N'-phenyl-, Urea, 1,1-dimethyl-3-phenyl- (8CI), N,N-Dimethyl-N'-phenylurea, 1,1-Dimethyl-3-phenylurea, 1-Phenyl-3,3-dimethylurea, 3-Phenyl-1,1-dimethylurea, Amicure UR, Dibar, Dybar, Dyhard 300, Dyhard RU 300, Dyhard UR 300, Falisilvan, Fenuron, Fikure 62U, N-Phenyl-N',N'-dimethylurea, Omicure 94, Omicure U 405, PUD, UR 300 |
| InChI Formula | InChI=1S/C9H12N2O/c1-11(2)9(12)10-8-6-4-3-5-7-8/h3-7H,1-2H3,(H,10,12) |
| SMILES | CN(C)C(=O)Nc1ccccc1 |
| IUPAC Name | 1,1-dimethyl-3-phenylurea |
| Molecular Weight (g/mol) | 164.2 |
| Formula Weight | 164.095 |