Learn More
Fenticonazole Nitrate, TRC
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Specifications
Specifications
| Chemical Name or Material | Fenticonazole nitrate |
| CAS | 73151-29-8 |
| Molecular Formula | C24 H20 Cl2 N2 O S . H N O3 |
| Quantity | 50 mg |
| Synonym | Fenticonazole nitrate, 1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[[4-(phenylsulfanyl)benzyl]oxy]ethyl]-1H-imidazole nitrate, 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-[[4-(phenylthio)phenyl]methoxy]ethyl]-, nitrate (1:1), 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-[[4-(phenylthio)phenyl]methoxy]ethyl]-, mononitrate (9CI), Falvin, Feniconazole nitrate, Fentiderm, REC 15-1476 |
| InChI Formula | InChI=1S/C24H20Cl2N2OS.HNO3/c25-19-8-11-22(23(26)14-19)24(15-28-13-12-27-17-28)29-16-18-6-9-21(10-7-18)30-20-4-2-1-3-5-20;2-1(3)4/h1-14,17,24H,15-16H2;(H,2,3,4) |
| SMILES | O[N+](=O)[O-].Clc1ccc(C(Cn2ccnc2)OCc3ccc(Sc4ccccc4)cc3)c(Cl)c1 |
| IUPAC Name | 1-[2-(2,4-dichlorophenyl)-2-[(4-phenylsulfanylphenyl)methoxy]ethyl]imidazole;nitric acid |
| Molecular Weight (g/mol) | 518.41 |
| Formula Weight | 517.063 |