Learn More
Fenoterol hydrobromide, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| CAS | 13392-18-2 |
| Color | White |
| Molecular Formula | C17H21NO4 |
| Quantity | 1 g |
| Synonym | fenoterol, phenoterol, fenoterolum, berotec, fenoterolum inn-latin, berotek, p-hydroxyphenylorciprenaline, p-hydroxyphenyl-orciprenaline, fenoterol usan:ban:inn, 1-3,5-dihydroxyphenyl-1-hydroxy-2-4-hydroxyphenyl isopropylamino ethane |
| InChI Key | LSLYOANBFKQKPT-UHFFFAOYSA-N |
| SMILES | CC(CC1=CC=C(C=C1)O)NCC(C2=CC(=CC(=C2)O)O)O |
| IUPAC Name | 5-[1-hydroxy-2-[1-(4-hydroxyphenyl)propan-2-ylamino]ethyl]benzene-1,3-diol |
| Molecular Weight (g/mol) | 303.358 |
| PubChem CID | 3343 |
| Show More |