Learn More
Fenbufen Ethyl Ester, Mikromol™
Description
Fenbufen Ethyl Ester
Specifications
Specifications
| Chemical Name or Material | Fenbufen Ethyl Ester |
| CAS | 1230-54-2 |
| Analyte or Component Names | Fenbufen Ethyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H18 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Fenbufen Ethyl Ester, gamma-oxo-[1,1'-Biphenyl]-4-butanoic acid ethyl ester, Propionic acid, 3-(4-biphenylylcarbonyl)-, ethyl ester (7CI), Ethyl 4-(4-phenylphenyl)-4-oxobutyrate, Ethyl gamma-oxo[1,1'-biphenyl]-4-butanoate |
| InChI Formula | InChI=1S/C18H18O3/c1-2-21-18(20)13-12-17(19)16-10-8-15(9-11-16)14-6-4-3-5-7-14/h3-11H,2,12-13H2,1H3 |
| SMILES | CCOC(=O)CCC(=O)c1ccc(cc1)c2ccccc2 |
| IUPAC Name | ethyl 4-oxo-4-(4-phenylphenyl)butanoate |
| Show More |