missing translation for 'onlineSavingsMsg'
Learn More

Fenbendazole, MedChemExpress

Product Code. p-200101781
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16442126

Brand: Medchem Express HYB0413

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fenbendazole is a broad spectrum benzimidazole anthelmintic used against gastrointestinal parasites.

Fenbendazole is a broad spectrum benzimidazole anthelmintic used against gastrointestinal parasites.

Specifications

Chemical Name or Material Fenbendazole
Physical Form Solid
CAS 43210-67-9
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C15H13N3O2S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 10 mg/mL (33.41 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(OC)NC1=NC2=CC=C(SC3=CC=CC=C3)C=C2N1
Molecular Weight (g/mol) 299.35
Formula Weight 299.35
Percent Purity 99.75%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.