Learn More
Value Plastics Female Luer Thread Style to UNF Threaded Adapter Fittings, Straight
Shop All Cole-Parmer Products
Click to view available options
Thread Size:
1/4-28 UNF(M)
10-32 UNF(M)
Inlet:
Female Luer with 1/4 in. Hex
Female Luer with 5/16 in. Hex
Material (Seal):
Nylon
Polycarbonate
Polypropylene
Unit Size:
Pack of 25
Description
Quick and easy assembly and disassembly without the need for tools. Thread design provides a secure closure—preventing contamination. Conforms to ISO-594-1 and ISO 594-2 standards
Specifications
Specifications
| Certifications/Compliance | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Packaging Quantity | 25/Pk. |
| Inlet | Female Luer with 5/16 in. Hex |
| Thread Size | 1/4-28 UNF(M) |
| Material (Seal) | Polypropylene |