Learn More
Value Plastics Female Luer Thread Style to NPT Adapter Fittings, Straight
Product Code. 30053952 Shop All Cole-Parmer Products
Change view
Click to view available options
Material (Seal):
Prolypropelene
White Nylon
Unit Size:
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material (Seal) Unit Size
30053952 Prolypropelene Pack of 25
30052983 White Nylon Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30053952 Supplier Value Plastics Supplier No. 18FTLL6005
  • ONLINE EXCLUSIVE PRICE
    802.00 NOK
    673.46 NOK / Pack of 25
    Save 128.54 NOK
    16% Off
Estimated Shipment: 19-01-2027
Luer fittings allow for a simple yet secure connection
Quick and easy assembly and disassembly without the need for tools. Thread design provides a secure closure—preventing contamination. Conforms to ISO-594-1 and ISO 594-2 standards
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Inlet 1/8-27 NPT(M) Thread
Material (Seal) Prolypropelene