Learn More
Cytiva Female Luer
Shop All Cytiva Products
Click to view available options
Connectors:
Female Luer to Female Luer
Female Luer to Male Luer
Unit Size:
Each
Pack of 10
Description
Polyamide connectors, pack of 10.Accessory for the Cytiva MidGee and MidGee Hoop cross-flow cartridge filtration system
Specifications
Specifications
| Connectors | Female Luer to Male Luer |
| For Use With (Equipment) | Male Luer Adopter |
| Material | Nylon |
| Type | Luer Adapter |