missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FEM1A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15169-1-AP-20UL
This item is not returnable.
View return policy
Description
FEM1A (FEM-1 homolog a), also known as EPRAP (Prostaglandin E receptor 4-associated protein), is a 669 amino acid cytoplasmic protein that belongs to the FEM-1 family. FEM1A is present in atheromata and in macrophages derived from peripheral blood monocytes. FEM1A may be a component of an E3 ubiquitin-protein ligase complex and functions as a substrate recognition subunit. It is suggested that FEM1A participates in anti-inflammatory signaling of macrophages via its interaction with EP4. FEM1A is down-regulated in Rhabdomyosarcoma, the most common soft tissue neoplasm of children. FEM1A is considered a candidate gene for polycystic ovary syndrome.Specifications
| FEM1A | |
| Polyclonal | |
| Unconjugated | |
| FEM1A | |
| EPRAP, fem 1 homolog a (C. elegans), FEM1 alpha, FEM1A, Protein fem 1 homolog A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55527 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BSK4 | |
| FEM1A | |
| FEM1A Fusion Protein Ag7357 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction