Description
FDPS Monoclonal specifically detects FDPS in Mouse samples. It is validated for ELISA.
Specifications
Specifications
| Classification | Monoclonal |
| Clone | 115 |
| Dilution | ELISA |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Mouse FDPS (Accession#: NP_608219.1; Met1-Lys353). |
| Quantity | 0.1 mL |
| Research Discipline | Core ESC Like Genes, Stem Cell Markers |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 2224 |
| Product Type | Antibody |
| Product Line | Novus |