Learn More
Ohaus™ FD Series Food Portioning Scale
Product Code. 11860152 Shop All Ohaus Products
Click to view available options
Model:
FD15
FD6
Unit Size:
Each
Product Code. 11860152
Brand: Ohaus™ 71170444
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A multi-use portioning scale designed for use in the food service industry
The OHAUS FD is a rugged, multi-use portioning scale designed for use in food service applications. Ideally suited for food portioning and checkweighing, the FD combines an easily cleaned design, fast display and high precision load cell. Designed for use in restaurants, bakeries, food manufacturing and other areas requiring fast operation and flexibility, the OHAUS FD scale offers durability and cleanliness in one economical design
  • All stainless steel construction
  • High resolution weighing cells with advanced electronics provide resolution up to 30,000d
  • NSF listed/certified to NSF/ANSI 169-2005 (formerly NSF C-2) and USDA-AMS accepted to NSF/3-A 14159-1, supports HACCP compliance
  • Check Weighing mode with 3 LED indicator and audible alarm
  • Bright backlit LCD display
  • Stable weight display within 2 seconds
  • Multiple weighing units including percent weighing, lb, decimal and fractional oz, kg, g, and lb:oz
  • 120-hour internal rechargeable battery or AC power
  • Industry leading warranty and support
TRUSTED_SUSTAINABILITY
Tekniske data
Battery Life 120 Hr.
Battery Type Internal rechargeable sealed lead-acid battery
Calibration User-selectable external span or linearity calibration
Certifications/Compliance NSF, HACCP
Construction Stainless Steel housing and platform, up-front level indicator
Capacity 6 kg
Display Type 5-digit, 7-segment Backlit Blue LCD
Readability 0.5 g
Keypad Type 4 button membrane switch
Linearity ±1 g
Model FD6
Protection Class IP43
Stabilization Time <2 sec.
Temperature (Metric) Operating -10°C to +40°C
Type FD Series Food Portioning Scale
Weighing Modes Pound, decimal ounce, 3/4, 1/2 and 1/4 fractional ounce, gram, kilogram, combined lb:oz display (either decimal or fractional ounces)
Width (Metric) 300 mm
Height (Metric) 223 mm
Depth (Metric) 71 mm
Weight (Metric) 3.6 kg
Electrical Requirements 9 VDC 0.5 A AC Wall Adapter, 100/120 VAC 50/60 Hz
Voltage 9/100 to 120 V
Frequency 50/60 Hz
Vis mere Vis mindre
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.