missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FCER1A (Human) Recombinant Protein
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
- Sequence: MAPAMESPTLLCVALLFFAPDGVLAVPQKPKVSLNPPWNRIFKGENVTLTCNGNNFFEVSSTKWFHNGSLSEETNSSLNIVNAKFEDSGEYKCQHQQVNESEPVYLEVFSDWLLLQASAEVVMEGQPLFLRCHGWRNWDVYKVIYYKDGEALKYWYENHNISITNATVEDSGTYYCTGKVWQLDYESEPLNITVIKAPREKYWLQFFIPLLVVILFAVDTGLFISTQQQVTFLLKIKRTRKGFRLLNPHPKPNPKNN
Specifications
Specifications
| Accession Number | AAH05912.1 |
| Gene ID (Entrez) | 2205 |
| Name | Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide |
| Preparation Method | Wheat germ expression system |
| Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
| Quantity | 10 μg |
| Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Gene Alias | FCE1A, FcERI |
| Gene Symbol | FCER1A |
| Species | Wheat Germ (in vitro) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur