missing translation for 'onlineSavingsMsg'
Learn More

Fc Receptor Binding Inhibitor Polyclonal Antibody, Functional Grade, eBioscience™, Invitrogen™

Product Code. 17111582
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
Product Code. Quantity unitSize
17111582 50 mg 50mg
17121582 5 mg 5mg
17101582 25 mg 25mg
3 options
This item is not returnable. View return policy

Product Code. 17111582

Brand: Affymetrix eBioscience 169161050

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Polyclonal Antibody

The human FcgammaR-binding inhibitor can be used to inhibit the non-specific Fc-gamma receptor (FcgammaR)-mediated binding of mouse monoclonal antibodies used for flow cytometric analysis of human tissue. Four different classes of Fcgamma receptors have been identified: FcgammaRI (CD64), FcgammaRII (CD32), and FcgammaRIII (CD16), and they are expressed at varying levels in multiple cell lineages including high expression in myeloid, granulocyte and B cells. The biological function of the FcgammaR, including initiation of endocytosis, phagocytosis and antigen presentation, is elicited upon binding of host-immunoglobulin. The extent to which mouse monoclonal antibodies will bind to human FcgammaR varies depending on the isotype of the MAb. Furthermore, different monoclonal antibodies of the same isotype will display different binding to human FcgammaRs.

Fc Receptor Binding Inhibitor Polyclonal antibody specifically detects Fc Receptor Binding Inhibitor in Human samples. It is validated for Control, Flow Cytometry, Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fc Receptor Binding Inhibitor
Applications Flow Cytometry, Functional Assay
Classification Polyclonal
Concentration 20 μL/Test
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2
Gene Alias Fc blocking; FcR BI; FcR Inhibitor
Purification Method Affinity chromatography
Quantity 50 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.