missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXW11 Mouse anti-Human, Mouse, Rat, Clone: 1F5A10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68090-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
FBXW11 Monoclonal antibody apecifically detetects FBXW11 antigen in Human, Mouse, Rat Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.Specifications
FBXW11 | |
Monoclonal | |
1000 μg/mL | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
FBXW11 | |
FBXW11 fusion protein Ag3761 | |
150 μL | |
23291 | |
Store at -20°C. Stable for one year after shipment. | |
IgG1 |
Western Blot, Immunohistochemistry, ELISA | |
1F5A10 | |
Unconjugated | |
BC026213 | |
Mouse | |
Protein A purification | |
Primary | |
Human, Mouse, Rat | |
Liquid |