Learn More
FBL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16895464 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16895464 20 μL 20µL
16885464 150 μL 150µL
2 options
Product Code. 16895464
Brand: Proteintech 160211AP20UL
Rabbit Polyclonal Antibody
Nop1p / Fibrillarin was originally identified as a nucleolar protein of bakers yeast, Saccharomyces cerevisiae (accession P15646). The Nop1p protein is essential for yeast viability and is localized in the nucleoli. The human homologue of Nop1p is fibrillarin (accession P22087) a component of the nucleolar small nuclear ribonucleoprotein (snRNP) particle. The human fibrillarin gene is located on chromosome 19 (19q13.1).
Specifications
Antigen FBL
Applications Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FBL
Gene Accession No. P22087, P22509, P35550
Gene Alias FBL, FIB, FIB1, fibrillarin, FLRN, RNU3IP1
Gene Symbols FBL
Host Species Rabbit
Immunogen FBL Fusion Protein Ag8841
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14113, 2091, 292747
Target Species Rat, Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG