missing translation for 'onlineSavingsMsg'
Learn More

FBF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11531-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16848673

  • 3828.53 NOK / 150µL
Estimated Shipment: 16-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is induced by N-terminal (20-347aa) of FBF1. It recognizes all the six isoforms.

Keratin-binding protein required for epithelial cell polarization. Involved in apical junction complex (AJC) assembly via its interaction with PARD3. Required for ciliogenesis.
Specifications

Specifications

FBF1
Polyclonal
Unconjugated
FBF1
Alb, albatross, Fas (TNFRSF6) binding factor 1, FBF 1, FBF1, FBF1 Ens, HCG1989313, isoform CRA_b, KIAA1863
Rabbit
Antigen Affinity Chromatography
RUO
217335, 85302
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A2A870, Q8TES7
FBF1
FBF1 Fusion Protein Ag2106
150 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers