Learn More
FB23-2, MedChemExpress
Product Code. 16448017
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. 16448017
Brand: Medchem Express HY127103
MedChemExpress FB23-2 is a potent and selective inhibitor of mRNA N6-methyladenosine (m6A) demethylase FTO, with an IC50 of 2.6 μM. FB23-2 has anti-proliferation activity. FB23-2 can be used for the research of acute myeloid leukemia (AML).
FB23-2 is a potent and selective inhibitor of mRNA N6-methyladenosine (m6A) demethylase FTO, with an IC50 of 2.6 μM. FB23-2 has anti-proliferation activity. FB23-2 can be used for the research of acute myeloid leukemia (AML).
Specifications
Chemical Name or Material FB23-2
Physical Form Solid
CAS 2243736-45-8
Color Light Nude
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C18H15Cl2N3O3
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 50 mg/mL (127.47 mM; Need ultrasonic)
SMILES O=C(NO)C1=CC=CC=C1NC2=C(Cl)C=C(C3=C(C)ON=C3C)C=C2Cl
Molecular Weight (g/mol) 392.24
Formula Weight 392.24
Percent Purity 98.55%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.