Learn More
Favipiravir, Mikromol™
Description
Favipiravir
Specifications
Specifications
| Chemical Name or Material | Favipiravir |
| CAS | 259793-96-9 |
| Analyte or Component Names | Favipiravir |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C5H4FN3O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 6-Fluoro-3,4-dihydro-3-oxo-2-pyrazinecarboxamide, 6-Fluoro-3,4-dihydro-3-oxo-pyrazinecarboxamide, 6-Fluoro-3-hydroxy-2-pyrazinecarboxamide, Favipiravir, T 705, T 705 (Pharmaceutical) |
| InChI Formula | InChI=1S/C5H4FN3O2/c6-2-1-8-5(11)3(9-2)4(7)10/h1H,(H2,7,10)(H,8,11) |
| SMILES | NC(=O)C1=NC(=CNC1=O)F |
| IUPAC Name | 5-fluoro-2-oxo-1H-pyrazine-3-carboxamide |
| Show More |