missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAT1 Antibody (FAT-1 3D7/1), Novus Biologicals™
Mouse Monoclonal Antibody
1740.00 NOK
Specifications
| Antigen | FAT1 |
|---|---|
| Clone | FAT-1 3D7/1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot |
Description
FAT1 Monoclonal specifically detects FAT1 in Drosophila samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| FAT1 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| FAT tumor suppressor 1, FAT tumor suppressor homolog 1 (Drosophila) | |
| FAT1 | |
| IgM | |
| Thiophilic Chromatography | |
| FAT (Fat-1). Tested only in Drosophila. Expected to cross-react with Rat (85% identity with immunogen) and Chimpanzee (92% identity with immunogen) due to sequence homology. |
| FAT-1 3D7/1 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Drosophila | |
| 2195 | |
| Cytoplasmic domain of drosophila Fat | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title