Learn More
Thermo Scientific™ FastVac™ Vacuum Manifold
Artikelnummer. 15822145 Alle ansehen Thermo Scientific Produkte
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 Ea.
missing translation for 'unitSize':
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity missing translation for 'unitSize'
15822145 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15822145 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' A35899
Black Friday Starts Here
Use promo code 27984
Alle Werbeaktionen anzeigen
Estimated Shipment: 02-12-2026
Zum Warenkorb hinzufügen
Edge
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
The one-way Luer-Lock stopcocks are compatible with most nucleic acid purification columns and individually controlled, allowing the processing of as few as one sample at a time.
The one-way Luer-Lock stopcocks are compatible with most nucleic acid purification columns and individually controlled, allowing the processing of as few as one sample at a time. Furthermore, the extra-large waste reservoir makes the FastVac Vacuum Manifold convenient for protocols that require the processing of large volumes, such as during plasmid DNA isolation using a PureLink Fast Low-Endotoxin plasmid purification kit.
Product features:
• Capacity—capable of processing 1 to 20 samples simultaneously using columns and a vacuum source
• Composition—consists of a linear, low-density polyethylene manifold body, one-way Luer-Lock stopcocks made of a polycarbonate body and polypropylene handle, a polypropylene vacuum hose connector, and a neoprene stopper
• Compatibility—compatible with most nucleic acid purification kits (refer to the kit instruction manual for information regarding compatibility and processing with a vacuum manifold)
TRUSTED_SUSTAINABILITY
Spezifikation
Product Type Vacuum Manifold
Content And Storage • 1 vacuum manifold
• 20 Luer-Lock connectors
• 1 Neoprene stopper
Product Line FastVac
Quantity 1 Ea.
Shipping Condition Room Temperature