missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fascin Recombinant Rabbit Monoclonal Antibody (1B3V6)
GREENER_CHOICE

Product Code. 17106473
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17106473 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17106473 Supplier Invitrogen™ Supplier No. MA535811

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fascin Recombinant Rabbit Monoclonal Antibody (ARC1638)

Fascin Recombinant Monoclonal Antibody for Western Blot

Fascin organizes filamentous actin into bundles with a minimum of 4. 1:1 actin/fascin ratio. It is probably involved in the assembly of actin filament bundles present in microspikes, membrane ruffles, and stress fibers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fascin
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 1B3V6
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FSCN1
Gene Accession No. P85845, Q16658, Q61553
Gene Alias 55 kDa actin-bundling protein; 55kDa actin-bundling protein; AI663989; Fan1; fascin; fascin 1; fascin actin-bundling protein 1; Fascin homolog 1; fascin homolog 1, actin bundling protein; fascin homolog 1, actin bundling protein (Strongylocentrotus purpuratus); fascin homolog 1, actin-bundling protein; Fascin1; fascin-1; Fscn1; fscn1 {ECO:0000250; HSN; p55; Singed; Singed (Drosophila) like (sea urchin fascin homolog like); singed-like (fascin homolog, sea urchin); singed-like protein; SNL; UniProtKB:Q16658}
Gene Symbols FSCN1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 249-362 of human Fascin/FSCN1 (Q16658).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14086, 6624, 683788
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.