Learn More
FARSB Mouse anti-Human, Unconjugated, Clone: 1H10C2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67924-1-IG-20UL
 View more versions of this product
Product Code. 17521436
  • 1710.33 NOK / 20µL
Description
The FARSB antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human FARSB. This antibody recognizes human antigen. The FARSB antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
FARSB
Monoclonal
Unconjugated
20 μL
10056
IgG2a
Western Blot, Immunohistochemistry
1H10C2