Learn More
FARSB Mouse anti-Human, Unconjugated, Clone: 1H10C2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67924-1-IG-150UL
Additional Details : Weight : 0.00020kg
Description
The FARSB antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human FARSB. This antibody recognizes human antigen. The FARSB antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
FARSB | |
Monoclonal | |
Unconjugated | |
150 μL | |
10056 | |
IgG2a |
Western Blot, Immunohistochemistry | |
1H10C2 | |
Mouse | |
Primary | |
Human |