Learn More
Farnesol, mixture of isomers, 96%
Product Code. 11439576
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11439576
Brand: Thermo Scientific Alfa Aesar A19316.22
Request Bulk or Custom Format
CAS: 4602-84-0 | C15H26O | 222.37 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
It can be used to induce apoptosis in cell cultures. It is also used as an antimicrobial agent. It is used in perfumes and as a flavoring agent.
Solubility
Not miscible or difficult to mix in water.
Notes
Light sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4602-84-0
Molecular Formula C15H26O
Molecular Weight (g/mol) 222.37
MDL Number MFCD00002918,MFCD00002918,MFCD00002918,MFCD00002918
InChI Key CRDAMVZIKSXKFV-YFVJMOTDSA-N
Synonym e,z-farnesol, 2-trans,6-cis-farnesol, 2e,6z-farnesol, trans,cis-farnesol, farnesol, 2e,6z-3,7,11-trimethyldodeca-2,6,10-trien-1-ol, farnesyl alcohol, e,z-3,7,11-trimethyl-2,6,10-dodecatrien-1-ol, 2-trans,6-cis-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
PubChem CID 1549109
ChEBI CHEBI:35966
IUPAC Name (2E,6Z)-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
SMILES CC(C)=CCC\C(C)=C\CC\C(C)=C\CO
Specifications
Density 0.887
Boiling Point 159°C to 160°C (10 mmHg)
Flash Point 146°C (295°F)
Odor Pleasant
Refractive Index 1.489
Quantity 100 g
Beilstein 1763926
Sensitivity Light sensitive
Merck Index 14,3937
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 222.37
Percent Purity 96%
Chemical Name or Material Farnesol, mixture of isomers