Learn More
Hellma™ Far UV Quartz Matched Pairs Macro Cells
Click to view available options
Capacity (Metric):
1.75 mL
Unit Size:
Pack of 2
Specifications
Specifications
| Type | Matched Pairs Macro Cell |
| Capacity (Metric) | 1.75 mL |
| Material | Far UV Quartz |
| Quantity | 2Pack |
| Path Length | 5 mm |
| Wavelength Range | 200 to 2500 nm |