missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAP Monoclonal Antibody (C1), Invitrogen™

Description
The protein encoded by this gene is a homodimeric integral membrane gelatinase belonging to the serine protease family. It is selectively expressed in reactive stromal fibroblasts of epithelial cancers, granulation tissue of healing wounds, and malignant cells of bone and soft tissue sarcomas. This protein is thought to be involved in the control of fibroblast growth or epithelial-mesenchymal interactions during development, tissue repair, and epithelial carcinogenesis.
Specifications
Specifications
| Antigen | FAP |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | C1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | FAP |
| Gene Accession No. | Q12884 |
| Gene Alias | 170 kDa melanoma membrane-bound gelatinase; Antiplasmin-cleaving enzyme FAP, soluble form; APCE; Dipeptidyl peptidase FAP; DPPIV; Fap; FAPA; FAPalpha; fibroblast activation protein; fibroblast activation protein alpha; fibroblast activation protein, alpha; gelatine degradation protease FAP; Integral membrane serine protease; post-proline cleaving enzyme; Prolyl endopeptidase FAP; seprase; Serine integral membrane protease; SIMP; Surface-expressed protease |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?