Specifications
| Antigen | FAM71F2 |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.66 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | FAM71F2 |
| Gene Accession No. | Q6NXP2 |
| Gene Alias | FAM137B, FAM71F2, Protein FAM137B, Protein FAM71F2 |
| Gene Symbols | FAM71F2 |
| Show More |