Learn More
FAM71F1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27984-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| FAM71F1 | |
| Polyclonal | |
| Unconjugated | |
| FAM71F1 | |
| FAM137A, FAM71F1, NYD SP18, Protein FAM137A, Protein FAM71F1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 84691 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96KD3 | |
| FAM71F1 | |
| FAM71F1 Fusion Protein Ag16525 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |