missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ FAM53A (Human) Recombinant Protein

Product Code. 16128743
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. Quantity unitSize
16128743 10 μg 10µg
16138743 25 μg 25µg
2 options
This item is not returnable. View return policy

Product Code. 16128743

Brand: Abnova™ H00152877P01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human FAM53A full-length ORF ( NP_001013644.1, 1 a.a. - 398 a.a.) recombinant protein with GST-tag at N-terminal.

  • Sequence: MVTLITEKLQSQSLDDLTCKAEAGPLQYSAETLNKSGRLFPLELNDQSPWKVFSGGPPVRSQAATGPDFSFLPGLSAAAHTMGLQWQPQSPRPGAGLGAASTVDPSESTGSSTAPPTKRHCRSLSEPEELVRCRSPWRPGSSKVWTPVSKRRCDSGGSATRQGSPGAVLPRSAVWSTGPTSPATPRPSSASGGFVDSSEGSAGSGPLWCSAESCLPSTRRRPSLSQERLAGAGTPLPWASSSPTSTPALGGRRGLLRCRSQPCVLSGKRSRRKRRREEDARWTRPSLDFLKMTQTLKNSKSLCSLNYEDDDEDDTPVKTVLSSPCDSRGLPGITMPGCSQRGLRTSPVHPNLWASRESVTSDGSRRSSGDPRDGDSVGEEGVFPRARWELDLEQIENN

Specifications

Accession Number NP_001013644.1
Gene ID (Entrez) 152877
Name family with sequence similarity 53, member A
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias DNTNP
Gene Symbol FAM53A
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.