missing translation for 'onlineSavingsMsg'
Learn More

FAM160B2 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 16846984 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16846984 150 μL 150µL
16856984 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16846984

Brand: Proteintech 171471AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FAM160B2, also named as RAI16, may play a role in early lens development in mammals and in upregulation of genes in CD34+ cells in coronary artery disease patients.

Specifications

Antigen FAM160B2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FAM160B2
Gene Accession No. Q86V87
Gene Alias FAM160B2, Protein FAM160B2, RAI16
Gene Symbols FAM160B2
Host Species Rabbit
Immunogen FAM160B2 Fusion Protein Ag10920
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64760
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.