missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM160B2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17147-1-AP-20UL
This item is not returnable.
View return policy
Description
FAM160B2, also named as RAI16, may play a role in early lens development in mammals and in upregulation of genes in CD34+ cells in coronary artery disease patients.Specifications
| FAM160B2 | |
| Polyclonal | |
| Unconjugated | |
| FAM160B2 | |
| FAM160B2, Protein FAM160B2, RAI16 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64760 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86V87 | |
| FAM160B2 | |
| FAM160B2 Fusion Protein Ag10920 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction