Learn More
Invitrogen™ FAM134C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114005
Description
Immunogen sequence: EAAQRALVEV LGPYEPLLSR VQAALVWERP ARSALWCLGL NAAFWFFALT For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 100%; Rat: 100%.
RETREG3 gene ontology annotations related to this gene include positive regulation of neuron projection development.
Specifications
| FAM134C | |
| Polyclonal | |
| Unconjugated | |
| RETREG3 | |
| 1300010M03Rik; 4933404C01Rik; AI551748; DKFZp686B1036; Fam134c; family with sequence similarity 134 member C; family with sequence similarity 134, member C; FLJ33806; protein FAM134C; Reticulophagy regulator 3; RETREG3; RGD1561189 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 162427 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q86VR2 | |
| RETREG3 | |
| Synthetic peptide directed towards the N terminal region of human FAM134C (aa 36-85). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.