missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM103A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19422-1-AP-150UL
This item is not returnable.
View return policy
Description
FAM103A1, also named as RNMT-activating mini protein, is a 118 amino acid protein, which belongs to the RAM family. FAM103A1 as a component of mRNA cap binding complex localizes in the nucleus. FAM103A1 is required for efficient mRNA cap methylation and regulates RNMT expression by a post-transcriptional stabilizing mechanism.Specifications
| FAM103A1 | |
| Polyclonal | |
| Unconjugated | |
| FAM103A1 | |
| C15orf18, FAM103A1, HsT19360, Protein FAM103A1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 293058, 67148, 83640 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BTL3, Q9CQY2 | |
| Fam103a1 | |
| FAM103A1 Fusion Protein Ag13749 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction