Learn More
Falcon™ Tissue Culture Treated Flasks
BIOPHARMA
Product Code. p-122715 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
70 mL
250 mL
600 mL
750 mL
800 mL
Culture Area:
12.5 cm2
150 cm2
175 cm2
225 cm2
25 cm²
25 cm2
75 cm²
75 cm2
Closure Type:
Plug Seal
Vented
Neck Style:
Canted
Straight
Unit Size:
Pack of 100
Pack of 200
Pack of 30
Pack of 40
Pack of 60
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Culture Area Closure Type Neck Style unitSize
10354331 25 mL 12.5 cm2 Plug Seal Canted Pack of 100
10249120 25 mL 12.5 cm2 Vented Canted Pack of 100
10634501 50 mL 25 cm2 Plug Seal Canted Pack of 200
10350391 50 mL 25 cm² Vented Canted Pack of 100
10341011 70 mL 25 cm2 Plug Seal Canted Pack of 200
10700052 70 mL 25 cm2 Vented Canted Pack of 100
10527161 250 mL 75 cm² Plug Seal Canted Pack of 60
10797791 250 mL 75 cm2 Plug Seal Straight Pack of 100
10537161 250 mL 75 cm2 Vented Canted Pack of 60
10508662 250 mL 75 cm2 Vented Straight Pack of 100
10728502 600 mL 150 cm2 Plug Seal Canted Pack of 40
10499032 600 mL 150 cm2 Vented Canted Pack of 40
10078780 750 mL 175 cm2 Plug Seal Straight Pack of 40
10218550 750 mL 175 cm2 Vented Straight Pack of 40
10446421 800 mL 225 cm2 Plug Seal Canted Pack of 30
10687762 800 mL 225 cm2 Vented Canted Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Product Code. 10354331 Supplier Falcon™ Supplier No. 353018
Only null left
Add to basket
Add to basket
Manufactured from optically clear virgin polystyrene for easy viewing of sample or cells. Falcon™ Tissue Culture Treated Flasks features vacuum-gas plasma tissue culture treatment to ensure consistent growth surface.
  • Secure stacking provides stability from accidental tips or spills
  • Nonpyrogenic tested to less than 0.1 EU/ mL
  • Volumetric graduations and writing patch
  • Vented caps incorporate a 0.2 μm hydrophobic membrane for optimal gas exchange
  • Growth area and volume are nominal
TRUSTED_SUSTAINABILITY
Specifications
Capacity (English) 0.8 oz.
Closure Color Blue
Closure Material Polyethylene
Capacity (Metric) 25 mL
Material Polystyrene
For Use With (Application) For enhanced cell attachment and growth
Neck Style Canted
No. per Case 100/Cs.
No. per Pack 10/Pk.
Shape Rectangular
Surface Coating TC-Treated
Sterility Sterile
Bottom Shape Rectangular
Closure Type Plug Seal
Culture Area 12.5 cm2
Surface Treatment Tissue Culture-treated
Show More Show Less
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.