Learn More
Falcon™ Round-Bottom Polystyrene Test Tubes
BIOPHARMAGREENER_CHOICE
Product Code. p-196920 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
5 mL
8 mL
14 mL
16 mL
19 mL
Closure Type:
Not Included
Screw Cap
Snap Cap
Sterility:
Non-sterile
Sterile
Packaging:
1000/Cs.
Bag of 125, 1000/Cs.
Bag of 25, 500/Cs.
Bulk, 1000/Cs.
Individually wrapped, 500/Cs.
Unit Size:
Pack of 1000
Pack of 500
Product Code. Capacity (Metric) Closure Type Sterility Packaging unitSize
10694331 19 mL Screw Cap Sterile Individually wrapped, 500/Cs. Pack of 500
12678475 16 mL Screw Cap Sterile Individually wrapped, 500/Cs. Pack of 500
10769861 16 mL Screw Cap Sterile Bag of 125, 1000/Cs. Pack of 1000
10186360 5 mL Not Included Non-sterile Bulk, 1000/Cs. Pack of 1000
10100151 5 mL Snap Cap Sterile Bag of 25, 500/Cs. Pack of 500
10252421 14 mL Snap Cap Sterile Bag of 25, 500/Cs. Pack of 500
10568531 14 mL Snap Cap Sterile Bag of 25, 500/Cs. Pack of 500
10186400 5 mL Snap Cap Sterile Bag of 125, 1000/Cs. Pack of 1000
10222901 8 mL Screw Cap Sterile Bag of 125, 1000/Cs. Pack of 1000
10481751 14 mL Snap Cap Sterile Bag of 125, 1000/Cs. Pack of 1000
10579511 5 mL Not Included Sterile 1000/Cs. Pack of 1000
10088710 5 mL Snap Cap Sterile Individually wrapped, 500/Cs. Pack of 500
10408851 14 mL Not Included Sterile Bag of 125, 1000/Cs. Pack of 1000
10769331 14 mL Snap Cap Sterile Individually wrapped, 500/Cs. Pack of 500
14 options
Product Code. 10694331
Brand: Falcon™ 352045
Only null left
Add to basket
Add to basket
Constructed of optically clear polystyrene for easy viewing of sample. Falcon™ Round-Bottom Polystyrene Tubes are widely used and referenced in laboratory protocols.
  • Widely used and referenced in laboratory protocols
  • Dual position snap-cap prevents sample loss with ease of handling
  • Sterile, nonpyrogenicity tested to less than 0.1 EU/mL (Cat. No. 352008 nonsterile)
The foundation for consistent research results
  • Provide reliable containment of laboratory fluid samples
  • Polystyrene tubes are best suited for procedures requiring high optical clarity
  • Dual-position snap caps, heavier gauge walls, and unique construction provide a secure, positive seal
  • Medical-style packaging materials
  • 1,400 RCF rating (RCF claims refer to Relative Centrifugal Force measured in g-force for materials with a specific gravity of 1.0, used in an appropriate rotor with correct cushion and safety precautions. Tubes used with organic solvents at temperatures below 0°C may have lower RCF ratings.)
  • Tube dimensions and volumes are approximate
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Black
Color Clear
Description 1400 x G
Capacity (Metric) 19 mL
Length (Metric) 150 mm
Material Polystyrene
Disposable Single-use
Includes Screw cap
Packaging Individually wrapped, 500/Cs.
Sterility Sterile
Graduated No
Diameter (Metric) 16 mm
Closure Type Screw Cap
Product Type Test Tube