missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14928-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes the last enzyme in the tyrosine catabolism pathway. FAH deficiency is associated with Type 1 hereditary tyrosinemia (HT).Specifications
FAH | |
Polyclonal | |
Unconjugated | |
FAH | |
Beta diketonase, FAA, FAH, Fumarylacetoacetase, Fumarylacetoacetate hydrolase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14085, 2184, 29383 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P16930, P25093, P35505 | |
Fah | |
FAH Fusion Protein Ag6704 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |