Learn More
FADS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10627-1-AP-20UL
 View more versions of this product
Product Code. 16857123
  • 1710.33 NOK / 20µL
Description
The protein encoded by this gene is a member of the fatty acid desaturase (FADS) gene family. Desaturase enzymes regulate unsaturation of fatty acids through the introduction of double bonds between defined carbons of the fatty acyl chain. FADS family members are considered fusion products composed of an N-terminal cytochrome b5-like domain and a C-terminal multiple membrane-spanning desaturase portion, both of which are characterized by conserved histidine motifs. This gene is clustered with family members FADS1 and FADS2 at 11q12-q13. 1; this cluster is thought to have arisen evolutionarily from gene duplication based on its similar exon/intron organization.
Specifications
Specifications
FADS1
Polyclonal
Unconjugated
FADS1
D5D, Delta(5) desaturase, Delta(5) fatty acid desaturase, FADS1, FADS6, FADSD5, fatty acid desaturase 1, LLCDL1, TU12
Rabbit
Antigen Affinity Chromatography
RUO
3992, 76267
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O60427, Q920L1
FADS1
FADS1 Fusion Protein Ag0978
20 μL
Primary
Mouse, Human