Learn More
FADS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
Antigen FADS
Concentration 0.21 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16852614
View Documents
Proteintech
14118-1-AP-20UL
20 μL
1710.33 NOK
20µL
View alternative products
Save Up to 
»
16842614
View Documents
Proteintech
14118-1-AP-150UL
150 μL
4836.10 NOK
150µL
View alternative products
Save Up to 
»
Description
Description
FLAD1 encodes the enzyme that catalyzes adenylation of flavin mononucleotide (FMN) to form flavin adenine dinucleotide (FAD) coenzyme.
Specifications
Specifications
FADS
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Rabbit
Rat, Human, Mouse
Q8NFF5, Q8R123
319945, 751787, 80308
FADS Fusion Protein Ag5267
Primary
-20°C
FLAD1
0.21 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
FAD pyrophosphorylase, FAD synthase, FAD1, FADS, FLAD1, FMN adenylyltransferase, PP591, RP11 307C12.7
Flad1
IgG
Antigen Affinity Chromatography