Learn More
Fadrozole Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Fadrozole Hydrochloride |
| CAS | 102676-31-3 |
| Molecular Formula | C14H13N3 . HCl |
| Quantity | 10 mg |
| Synonym | Imidazo[1,5-a]pyridine, benzonitrile deriv., Afema, CGS 16949, CGS 16949A, 4-(5,6,7,8-tetrahydroimidazo[1,5-a]pyridin-5-yl)benzonitrile |
| InChI Formula | InChI=1S/C14H13N3.ClH/c15-8-11-4-6-12(7-5-11)14-3-1-2-13-9-16-10-17(13)14;/h4-7,9-10,14H,1-3H2;1H |
| SMILES | Cl.N#Cc1ccc(cc1)C2CCCc3cncn23 |
| Molecular Weight (g/mol) | 259.73 |
| Formula Weight | 259.0876 |