Learn More
Factor VIII Antibody - BSA Free, Novus Biologicals™
Product Code. 18727803 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
Product Code. Quantity unitSize
18727803 0.1 mg 0.1mg
18787073 0.025 mg 25µg
2 options
Product Code. 18727803
Brand: Novus Biologicals NB10091761
Rabbit Polyclonal Antibody has been used in 4 publications
Factor VIII Polyclonal specifically detects Factor VIII in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Antigen Factor VIII
Applications Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)
Gene Symbols F8
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of human Factor VIII (between amino acids 2100-2250) [UniProt P00451]
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Target Species Mouse, Rat
For Research Use Only