Description
- Extended facial protection: Adds an extra layer of safety by protecting the lower face from impacts, particles, and debris.
- Designed for SPECTRUM compatibility: Specifically developed for seamless integration with SPECTRUM safety goggles (regular size).
- Improved coverage: Enhances overall PPE protection by extending the protective area beyond the eyes.
- Maintains comfort: Lightweight design ensures user comfort without compromising protection during extended wear.
- Secure and stable fit: Attaches firmly to goggles to remain in place during movement and demanding tasks.
- Supports compliance: Increased protection and comfort encourage consistent use in high-risk environments.
- Durable construction: Built to withstand daily industrial use, helping reduce replacement frequency.
- Ideal for high-exposure environments: Suitable for industries such as construction, manufacturing, and maintenance where additional face protection is required.
Specifications
Specifications
| Certifications/Compliance | EN ISO 16321-1 |
| Lens Coating | Uncoated |
| Color | Black |
| Lens Tint | Clear |
| Product Type | Safety Goggles |
| Frame Color | Blue |
| Material | Polycarbonate |
| Disposable | Reusable |
| Lens Material | Plastic |
| Frame Material | Polycarbonate |
| Show More |